PA1-26581 detects MCM3 from human samples. PA1-26581 is expected to cross react with mouse (98% conserved) due to sequence homology.
PA1-26581 has been successfully used in Western blot procedures.
The PA1-26581 immunogen is a synthetic peptide corresponding to a portion of human Minichromosome Maintenance Deficient 3 encoded in part by exons 1 and 2.
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein is a subunit of the protein complex that consists of MCM2-7. It has been shown to interact directly with MCM5/CDC46. This protein also interacts with, and thus is acetlyated by MCM3AP, a chromatin-associated acetyltransferase. The acetylation of this protein inhibits the initiation of DNA replication and cell cycle progression.