Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • MALT1 Antibodies

          Abnova

          MALT1 Monoclonal Antibody (1E12)

          View all (42) MALT1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MALT1 Monoclonal Antibody (1E12)

          • Antibody Testing Data (1)
          MALT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MALT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MALT1 Antibody (H00010892-M01) in WB

          Western Blot analysis of MALT1 expression in transfected 293T cell line by MALT1 monoclonal antibody (M01), clone 1E12. Lane 1: MALT1 transfected lysate (Predicted MW: 91.1 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MALT1 Antibody in Western Blot (WB)

          Product Details

          H00010892-M01

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1E12

          Immunogen

          MALT1 (NP_776216.1, 1 a.a. approximately 813 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.43 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          Immunogen sequence: MSLLGDPLQA LPPSAAPTGP LLAPPAGATL NRLREPLLRR LSELLDQAPE GRGWRRLAEL AGSRGRLRLS CLDLEQCSLK VLEPEGSPSL CLLKLMGEKG CTVTELSDFL QAMEHTEVLQ LLSPPGIKIT VNPESKAVLA GQFVKLCCRA TGHPFVQYQW FKMNKEIPNG NTSELIFNAV HVKDAGFYVC RVNNNFTFEF SQWSQLDVCD IPESFQRSVD GVSESKLQIC VEPTSQKLMP GSTLVLQCVA VGSPIPHYQW FKNELPLTHE TKKLYMVPYV DLEHQGTYWC HVYNDRDSQD SKKVEIIIDE LNNLGHPDNK EQTTDQPLAK DKVALLIGNM NYREHPKLKA PLVDVYELTN LLRQLDFKVV SLLDLTEYEM RNAVDEFLLL LDKGVYGLLY YAGHGYENFG NSFMVPVDAP NPYRSENCLC VQNILKLMQE KETGLNVFLL DMCRKRNDYD DTIPILDALK VTANIVFGYA TCQGAEAFEI QHSGLANGIF MKFLKDRLLE DKKITVLLDE VAEDMGKCHL TKGKQALEIR SSLSEKRALT DPIQGTEYSA ESLVRNLQWA KAHELPESMC LKFDCGVQIQ LGFAAEFSNV MIIYTSIVYK PPEIIMCDAY VTDFPLDLDI DPKDANKGTP EETGSYLVSK DLPKHCLYTR LSSLQKLKEH LVFTVCLSYQ YSGLEDTVED KQEVNVGKPL IAKLDMHRGL GRKTCFQTCL MSNGPYQSSA ATSGGAGHYH SLQDPFHGVY HSHPGNPSNV TPADSCHCSR TPDAFISSFA HHASCHFSRS NVPVETTDEI PFSFSDRLRI SEK

          Target Information

          MALT1/Paracaspase (MALT1) was independently identified as a member of the human paracaspase family and an interacting partner of B-cell lymphoma (Uren et al., 2000). MALT1 is a caspase-like protein that contains an N-terminal death domain, two Ig-like domains, and a C-terminal caspase-like domain. It binds to Bcl10 through its Ig-like domains and cooperates with Bcl10 to activate NF-kappaB.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: caspase-like protein; DKFZp434L132; MALT lymphoma-associated translocation; malt-1; MALT1 protease; mucosa associated lymphoid tissue lymphoma translocation gene 1; Mucosa-associated lymphoid tissue lymphoma translocation protein 1; Paracaspase; paracaspase 1; paracaspase-1

          View more View less

          Gene Aliases: IMD12; MALT1; MLT; MLT1; PCASP1

          View more View less

          UniProt ID: (Human) Q9UDY8

          View more View less

          Entrez Gene ID: (Human) 10892

          View more View less

          Function(s)
          protease binding cysteine-type endopeptidase activity ubiquitin-protein transferase activity signal transducer activity protein binding peptidase activity kinase activator activity protein self-association
          Process(es)
          B-1 B cell differentiation stimulatory C-type lectin receptor signaling pathway response to molecule of bacterial origin positive regulation of T cell cytokine production proteolysis defense response activation of NF-kappaB-inducing kinase activity response to fungus protein ubiquitination positive regulation of protein ubiquitination positive regulation of interleukin-2 production Fc-epsilon receptor signaling pathway T cell proliferation regulation of apoptotic process negative regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling innate immune response T cell receptor signaling pathway regulation of T cell receptor signaling pathway positive regulation of T cell activation positive regulation of NF-kappaB transcription factor activity nuclear export protein oligomerization
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66966984d8-2vwff:80/100.66.134.75:80.
          git-commit: 4f7892df6eac4c3aaa23131f1084e28be0ddeb54
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0