Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SERSHGHSER SHGHSERSHG HSKRSRSQGD LVDTQSDLIA TQRDLIATQK DLIATQRDLI ATQRDLIVTQ RDLVATERDL INQSGRSHGQ SERHQRYSTG K
Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 25%.
Export adapter involved in mRNA nuclear export in cancer cells. Binds and enhances the RNA-binding activity of the nuclear RNA export factor NXF1. Can restore mRNA export function in cells compromised by loss of mRNA export adapters.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 28; CT-28; HOM-TES-85 tumor antigen; Leucine zipper protein 4; Tumor antigen HOM-TES-85
Gene Aliases: CT-28; CT-8; CT28; HOM-TES-85; LUZP4
UniProt ID: (Human) Q9P127
Entrez Gene ID: (Human) 51213
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support