Keep as concentrated solution.
Predicted reactivity: Mouse (84%), Rat (86%), Pig (88%), Bovine (89%).
Positive Control: SK-N-SH, mouse kidney.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain. Mutations in LRRK2 have been associated with Parkinson's disease. LRRK2 is a member of the leucine-rich repeat kinase family and encodes a protein with an ankryin repeat region, a leucine-rich repeat domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain, and a WD40 domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane.