Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: HNQLRVLPPE VGKLTRIVVL NLCGNRLKSL PREVSLLQCL KVLFVSMNCL TEVPAELSLC RKLEVLSLSH NCLSQ
Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
LRRC30 (Leucine Rich Repeat Containing 30) is a Protein Coding gene. An important paralog of this gene is SHOC2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leucine-rich repeat-containing protein 30
Gene Aliases: LRRC30
UniProt ID: (Human) A6NM36
Entrez Gene ID: (Human) 339291
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support