Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: KMYGPGGGKY FSTTEDYDHE ITGLRVSVGL LLVKSVQVKL GDSWDVKLGA LGGNTQEVTL QPGEYITKVF VAFQAFLRGM VMYTSKDRYF YFGKLDGQIS SAYPSQEGQV LVGIYGQYQL LGIKSIGFEW NYPLEEPTTE PPVNLTYSAN SPVGR
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: endocrine and exocrine protein; jacalin-like lectin domain containing 2; Pancreatic adenocarcinoma up-regulated factor; pancreatic adenocarcinoma upregulated factor; Zymogen granule protein 16 homolog B
Gene Aliases: EECP; HRPE773; JCLN2; PAUF; PRO1567; UNQ773/PRO1567; ZG16B
UniProt ID: (Human) Q96DA0
Entrez Gene ID: (Human) 124220
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support