This antibody is predicted to react with mouse and rat based on sequence homology.
As part of the NSL complex it is involved in acetylation of nucleosomal histone H4 on several lysine residues and therefore may be involved in the regulation of transcription.