Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EEKREEGDTE PLLKQEGPVE AAQPPVERKE AAALEQGRKV EELRWQEVDE RQTMPRPYTF QVSSGGKQIL FPKVNLSPVT PAKDTGLTAA PQEPKAPKAS PVQ
Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 53%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer-related regulator of actin dynamics; Capping protein-inhibiting regulator of actin dynamics
Gene Aliases: CRACD; CRAD; KIAA1211
UniProt ID: (Human) Q6ZU35
Entrez Gene ID: (Human) 57482
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support