Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • JAK3 Antibodies

          Abnova

          JAK3 Monoclonal Antibody (2A5)

          View all (39) JAK3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite JAK3 Monoclonal Antibody (2A5)

          Product Details

          H00003718-M03

          Applications
          Tested Dilution
          Publications

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2A5

          Immunogen

          JAK3 (AAH28068, 1 a.a. approximately 619 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.3 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MAPPSEETPL IPQRSCSLLS TEAGALHVLL PARGPGPPQR LSFSFGDHLA EDLCVQAAKA SGILPVYHSL FALATEDLSC WFPPSHIFSV EDASTQVLLY RIRFYFPNWF GLEKCHRFGL RKDLASAILD LPVLEHLFAQ HRSDLVSGRL PVGLSLKEQG ECLSLAVLDL ARMAREQAQR PGELLKTVSY KACLPPSLRD LIQGLSFVTR RRIRRTVRRA LRRVAACQAD RHSLMAKYIM DLERLDPAGA AETFHVGLPG ALGGHDGLGL LRVAGDGGIA WTQGEQEVLQ PFCDFPEIVD ISIKQAPRVG PAGEHRLVTV TRTDNQILEA EFPGLPEALS FVALVDGYFR LTTDSQHFFC KEVAPPRLLE EVAEQCHGPI TLDFAINKLK TGGSRPGSYV LRRSPQDFDS FLLTVCVQNP LGPDYKGCLI RRSPTGTFLL VGLSRPHSSL RELLATCWDG GLHVDGVAVT LTSCCIPRPK EKSNLIVVQR GHSPPTSSLV QPQSQYQLSQ MTFHKIPADS LEWHENLGHG SFTKIYRGCR HEVVDGEARK TEVLLKVMDA KHKNCMESFL EAASLMSQVS YRHLVLLHGV CMAGDSESPP PTHPTPASPK SRLFFPPLF

          Target Information

          Janus kinase 3 (JAK3) phosphorylates members of the STAT (signal transduction and activators of transcription) family. JAK3 is predominantly expressed in immune cells and transduces a signal in response to activation of interleukin receptors. In response to a varitety of cytokine or related factors (e.g., interferon, interleukins), JAKs are activated via phosphorylation at 2 adjacent tyrosine residues. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. However, active JAK1 may be required for complex responses. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Breast-JAK3; JAK-3; JAK3B; Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); L JAK; L-JAK; Leukocyte janus kinase; Tyrosine-protein kinase JAK3

          View more View less

          Gene Aliases: JAK-3; JAK3; JAK3_HUMAN; JAKL; L-JAK; LJAK

          View more View less

          UniProt ID: (Human) P52333

          View more View less

          Entrez Gene ID: (Human) 3718

          View more View less

          Function(s)
          protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity Ras guanyl-nucleotide exchange factor activity receptor binding protein binding ATP binding protein phosphatase binding
          Process(es)
          MAPK cascade adaptive immune response negative regulation of dendritic cell cytokine production protein phosphorylation enzyme linked receptor protein signaling pathway positive regulation of cytosolic calcium ion concentration tyrosine phosphorylation of STAT protein STAT protein import into nucleus cell migration peptidyl-tyrosine phosphorylation B cell differentiation negative regulation of interleukin-10 production negative regulation of interleukin-12 production intracellular signal transduction interleukin-4-mediated signaling pathway peptidyl-tyrosine autophosphorylation positive regulation of T cell proliferation positive regulation of activated T cell proliferation tyrosine phosphorylation of Stat5 protein T cell homeostasis positive regulation of GTPase activity innate immune response negative regulation of FasL biosynthetic process negative regulation of T-helper 1 cell differentiation positive regulation of immune response negative regulation of T cell activation positive regulation of calcium ion transport JAK-STAT cascade involved in growth hormone signaling pathway regulation of T cell apoptotic process negative regulation of thymocyte apoptotic process response to interleukin-2 response to interleukin-4 response to interleukin-15 response to interleukin-9
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0