Search Thermo Fisher Scientific
Rockland
FIGURE: 1 / 1
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Reacts with a 14 -16 kDa protein as demonstrated by western blot analysis on the Raji cell line. This antigen was detected after 12h of PHA-induced activation in the early G1 phase but absent in non-stimulated lymphocytes. Cross reactivity is likely to occur with mouse and rat. Reactivity with other sources has not been determined.
IPO-38 antigen is present in the nuclei of proliferating cells such as Hodgkin's disease and non-Hodgkin's lymphomas, different forms of leukemias, breast and colorectal carcinomas, and PHA-stimulated lymphocytes. It is not expressed in the cells of non-stimulated lymphocytes and granulocytes. IPO-38 can be a useful marker of cell proliferation during monitoring of tumor progression.