PA1-28315 detects ICA1L from human samples.
PA1-28315 has been successfully used in Western blot applications.
The PA1-28315 immunogen is: Fusion protein: levfhsvqetctellkiiekyqlrlnviseeenelglflkfqaerdatqagkmmdatgkalcssakqrlalctplsrlkqevatfsqravsdtlmt, corresponding to amino acids 53-148 of Human Ica69-related protein/LOC130026.
ICA1L is a protein coding gene and gene ontology (GO) annotation includes acrosomal vesicle; protein domain specific binding; spermatid development.