Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • HSP90 Antibodies

          QED Bioscience

          Hsp90 Monoclonal Antibody (4F3.E8)

          View all (40) HSP90 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Hsp90 Monoclonal Antibody (4F3.E8)

          Product Details

          11113-100UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5-1 µg/mL
          -

          Immunohistochemistry (IHC)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          4F3.E8

          Immunogen

          Recombinant human Hsp90 purified from E. coli.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1.0 mg/mL

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1 and mitochondria (Chen et al., 2005 ).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; Heat shock 84 kDa; Heat shock 86 kDa; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein 1, beta; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; heat shock 90kDa protein 1, beta; Heat shock protein; heat shock protein 1, alpha; heat shock protein 1, beta; heat shock protein 86; heat shock protein 90 kDa; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein 90kDa alpha family class B member 1; Heat shock protein family C member 1; Heat shock protein family C member 3; Heat shock protein HSP 90-alpha; Heat shock protein HSP 90-beta; heat shock protein, 1; heat shock protein, 84 kDa 1; heat shock protein, 86 kDa 1; heat shock protein, 89 kDa; HSP; HSP 84; HSP 86; HSP 90; HSP84; HSP86; HSP90-beta; LAP-2; Lipopolysaccharide-associated protein 2; LPS-associated protein 2; Renal carcinoma antigen NY-REN-38; retinal degeneration slow protein; TSTA; Tumor-specific transplantation 84 kDa antigen; Tumor-specific transplantation 86 kDa antigen

          View more View less

          Gene Aliases: 86kDa; 89kDa; 90kDa; AL022974; AL024080; AL024147; C81438; D6S182; EL52; HEL-S-65p; hsp4; HSP84; Hsp84-1; HSP86; Hsp86-1; Hsp89; HSP89A; Hsp90; HSP90-BETA; HSP90A; HSP90AA1; HSP90AB1; HSP90B; HSP90N; HSPC1; HSPC2; HSPC3; HSPCA; HSPCAL1; HSPCAL4; HSPCB; HSPN; LAP-2; LAP2

          View more View less

          UniProt ID: (Human) P07900, (Human) P08238, (Mouse) P07901, (Rat) P82995, (Mouse) P11499, (Rat) P34058

          View more View less

          Entrez Gene ID: (Human) 3320, (Human) 3326, (Mouse) 15519, (Rat) 299331, (Mouse) 15516, (Rat) 301252

          View more View less

          Function(s)
          nucleotide binding UTP binding CTP binding mRNA binding protein tyrosine kinase activity protein binding ATP binding GTP binding ATPase activity sulfonylurea receptor binding protein kinase binding protein phosphatase binding MHC class II protein complex binding nitric-oxide synthase regulator activity TPR domain binding dATP binding identical protein binding protein homodimerization activity histone deacetylase binding ion channel binding poly(A) RNA binding GTPase binding Rho GDP-dissociation inhibitor binding unfolded protein binding double-stranded RNA binding protein kinase regulator activity kinase binding cadherin binding involved in cell-cell adhesion
          Process(es)
          placenta development protein folding response to stress virion attachment to host cell positive regulation of protein binding positive regulation of protein import into nucleus, translocation negative regulation of neuron apoptotic process positive regulation of cell size cellular response to interleukin-4 positive regulation of protein serine/threonine kinase activity fibril organization G2/M transition of mitotic cell cycle neuron migration skeletal muscle contraction mitochondrial transport receptor-mediated endocytosis response to unfolded protein signal transduction response to heat response to cold response to salt stress positive regulation of lamellipodium assembly cardiac muscle cell apoptotic process peptidyl-tyrosine phosphorylation regulation of protein ubiquitination Fc-gamma receptor signaling pathway involved in phagocytosis ERBB2 signaling pathway protein refolding regulation of protein complex assembly protein unfolding response to estrogen protein import into mitochondrial outer membrane positive regulation of nitric oxide biosynthetic process response to antibiotic vascular endothelial growth factor receptor signaling pathway protein stabilization regulation of nitric-oxide synthase activity chaperone-mediated protein complex assembly positive regulation of cardiac muscle contraction chaperone-mediated autophagy regulation of cellular response to heat nitric oxide biosynthetic process positive regulation of cytotoxic T cell differentiation regulation of catalytic activity negative regulation of proteasomal ubiquitin-dependent protein catabolic process regulation of interferon-gamma-mediated signaling pathway regulation of type I interferon-mediated signaling pathway cellular response to organic cyclic compound positive regulation of phosphoprotein phosphatase activity cell-cell adhesion
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66966984d8-2vwff:80/100.66.134.75:80.
          git-commit: 4f7892df6eac4c3aaa23131f1084e28be0ddeb54
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0