Search Thermo Fisher Scientific
Nordic
Reconstituted by adding 0.5 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20°C.
Cross-reactivities against enzymes of other sources may occur but have not been determined.
Prior to use, an aliquot is thawed slowly at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4°C, not refrozen, and preferably used t he same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the product.
Arylsulfatase is a type of sulfatase enzyme that facilitates the synthesis of thiohydroximates. It causes the hydrolysis of beta-naphthyl sulfate much faster than alpha-naphthyl sulfate and possesses regiospecificity in the hydrolysis of ortho and para substituted phenyl sulfates.