Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEELLPDGQI WANMDPEERM LAAATAFTHI CAGQGEGDVR REAQSIQYDP YSKASVAPGK RPALPVQLQY PHVESNVPSE TVSEASQRLR KPVMK
Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.
This gene encodes a protein localized to the cytoplasm. Mutations in this gene are associated with hydrolethalus syndrome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Centriolar and ciliogenesis-associated protein HYLS1; hydrolethalus syndrome 1; Hydrolethalus syndrome protein 1
Gene Aliases: HLS; HYLS1
UniProt ID: (Human) Q96M11
Entrez Gene ID: (Human) 219844
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support