Immunogen sequence: VYHHHHHHPY VHPQAPVAAA APDGRYMRSW LEPTPGALSF AGLPSSRPYG IKPEPLSARR GDCPTLDTHT LSLTDYACGS PPVD
HOXA9 is the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development.