Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
HNRNPUL2 gene ontology annotations related to this gene include RNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: heterogeneous nuclear ribonucleoprotein U-like 2; Heterogeneous nuclear ribonucleoprotein U-like protein 2; MLF1-associated nuclear protein; SAF-A2; Scaffold-attachment factor A2
Gene Aliases: 1110031M08Rik; AI465155; HNRNPUL2; HNRPUL2; Manp; SAF-A2
UniProt ID: (Human) Q1KMD3, (Mouse) Q00PI9
Entrez Gene ID: (Human) 221092, (Mouse) 68693
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support