PA1-32313 detects HLTF from human samples.
PA1-32313 has been successfully used in immunoprecipitation, Western blot applications.
The PA1-32313 immunogen is: Synthetic peptide of the human HLTF protein, amino acids 50-292.
This gene encodes a member of the SWI/SNF family. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein contains a RING finger DNA binding motif. Two transcript variants encoding the same protein have been found for this gene. However, use of an alternative translation start site produces an isoform that is truncated at the N-terminus compared to the full-length protein.