PA1-31579 detects SMARCA3 from human samples.
PA1-31579 has been successfully used in Western blot applications.
The PA1-31579 immunogen is: Peptide with sequence KQAKINEIRTLIDL, from C Terminus of the protein sequence according to NP_003062; NP_620636
This gene encodes a member of the SWI/SNF family. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein contains a RING finger DNA binding motif. Two transcript variants encoding the same protein have been found for this gene. However, use of an alternative translation start site produces an isoform that is truncated at the N-terminus compared to the full-length protein.