Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAADTPGKPS ASPMAGAPAS ASRTPDKPRS AAEHRKSSKP VMEKRRRARI NESLAQLKTL ILDALRKESS RHSKLEKADI LEMTVRHLRS LRRVQVTAAL SADPAVLGKY RAGFHECLAE VNRFLAGCEG VPADVRSRLL GHLAACLRQL GPSRRPASLS PAAPAEAPAP EVYAGRPLLP SLGGPFPLLA PPLLPGLTRA LPAAPRAGPQ GPGGPWRPWL R
HES4 is a protein coding gene. Transcriptional repressor. Binds DNA on N-box motifs: 5'-CACNAG-3'.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLH factor Hes4; bHLHb42; Class B basic helix-loop-helix protein 42; Hairy and enhancer of split 4; hHES4; Transcription factor HES-4
Gene Aliases: BHLHB42; HES4
UniProt ID: (Human) Q9HCC6
Entrez Gene ID: (Human) 57801
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support