PA5-29909 targets Granulins in IF, IHC (P), and WB applications and shows reactivity with Human samples.
The PA5-29909 immunogen is recombinant fragment corresponding to a region within amino acids 62 and 402 of Human Granulins.
PCDGF is a glycoprotein and growth factor involved in the regulation of mitosis, early embryonic development, cell division, cell migration, cell survival, and hypothalamic differentiation. PCDGF is also known as progranulin because it is the precursor for smaller proteins known as epithelins or granulins.