Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Glra4 is a member of glycine receptors (GlyR), a cys-loop family of ligand-gated ion channels responsible for mediating the inhibitory effects of glycine. GlyR are widely distributed throughout the CNS, particularly within the hippocampus, spinal cord and brain stem. Glycine receptors are ligand-gated chloride channels. Channel opening is triggered by extracellular glycine and channel opening is also triggered by taurine and beta-alanine. Glra4 plays a role in the down-regulation of neuronal excitability, and contributes to the generation of inhibitory postsynaptic currents. The Glra4 gene encodes a protein which has a neurotransmitter-gated ion-channel ligand binding domain. Glra4 protein is very similar to a mouse protein which is a subunit of the retinal glycine receptor. Multiple transcript variants encoding different isoforms have been found for the Glra4 gene. Diseases associated with GLRA4 include Pelizaeus-Merzbacher Disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glycine receptor subunit alpha-4; glycine receptor, alpha 4 subunit
Gene Aliases: Glra4; RGD1564103
UniProt ID: (Mouse) Q8VHF3
Entrez Gene ID: (Human) 441509, (Mouse) 14657, (Rat) 367912
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support