PA1-28258 detects Ghrelin from human, rat, mouse samples.
PA1-28258 has been successfully used in Western blot applications.
The PA1-28258 immunogen is: RKESKKPPAKLQ, corresponding to amino acids 38-49 of Ghrelin.
GHRL encodes ghrelin-obestatin preproprotein, which generates ghrelin and obestatin. Ghrelin is an the ligand for the growth hormone secretagogue receptor type 1 and is involved in regulating growth hormone release. Ghrelin has an appeptite-stimulating effect and induces adiposity and stimulates gastric acid secretion. Obestatin may be the ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and can have an appetite-reducing effect resulting in decreased food intake.