FIGURE: 1 / 4
Keep as concentrated solution.
Predicted reactivity: Mouse (93%), Rat (90%), Chicken (80%), Sheep (96%), Bovine (95%).
Positive Control: HeLa, mouse plasma, rat plasma.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene binds to the "plus" ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.