PA1-24494 detects GTF2H1 in Human samples.
PA1-24494 has been successfully used in Immunohistochemistry and Western Blot procedures.
PA1-24494 immunogen corresponds synthetic peptide corresponding to a region of human GTF2H1.
GTF2H1 (general transcription factor IIH, polypeptide 1, 62kDa) is a component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA. When complexed to CAK, GTF2H1 is involved in RNA transcription by RNA polymerase II.