Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: RIHQLALTGE KRADIQLNSF GFYTNGSLEV ELSVLRLGLR EAEEKSLLVG FSLSRVRSGR VRSYSTRDFQ DCPLQKNSSS FLVLFLINTK DLQVQVRKYG EQKTLFIFPG LLPEAPSKPG LPKPQATVPR KVDGGGTSAA SKPKSTPAVI QGPSGKDKDL VLGLSHLNNS YNFSFHVVIG SQAEEGQYSL NFHNCNNSVP GKEHPFDITV MIREKNPDGF LSAAEMPL; Positive Samples: U-251MG, SW620, Raji, Mouse liver, Mouse brain, Mouse lung, Rat heart; Cellular Location: Membrane, Multi-pass membrane protein
GPR108 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lung seven transmembrane receptor 2; Protein GPR108
Gene Aliases: 1810015L19Rik; AA589464; C79132; GPR108; LUSTR2
UniProt ID: (Human) Q9NPR9, (Rat) Q6P6V6, (Mouse) Q91WD0
Entrez Gene ID: (Human) 56927, (Rat) 316136, (Mouse) 78308
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support