Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total GNAO1.
Heterotrimeric G proteins are membrane bound GTPases that are linked to 7-TM receptors. Each G protein contains an alpha-, beta- and gamma-subunit and is bound to GDP in the 'off' state. Ligand-receptor binding results in detachment of the G protein, switching it to an 'on' state and permitting Galpha activation of second messenger signalling cascades. There are several types of Galpha proteins; Galphas and Galphai bind directly to adenylyl cyclase and stimulate or inhibit its activity, leading to an increase or decrease in cAMP levels respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686O0962; G alpha o; GNAO 1; Go alpha; GO2-q chimeric G-protein; GTP-binding protein alpha o; GTP-binding protein alpha-o subunit; guanine nucleotide binding protein (G protein), alpha activating activity polypeptide O; guanine nucleotide binding protein, alpha O polypeptide 1; Guanine nucleotide-binding protein G(o) subunit alpha; guanine nucleotide-binding regulatory protein 2
Gene Aliases: alphaO; AW050213; EIEE17; G-ALPHA-o; Galphao; Gna0; GNAO; GNAO1; HLA-DQB1; RATBPGTPC
UniProt ID: (Human) P09471, (Rat) P59215, (Mouse) P18872
Entrez Gene ID: (Human) 2775, (Rat) 50664, (Mouse) 14681
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support