PA1-85445 has exhibited the following cross reactivities: Human FSH - 66%, Rat LH - 8%, Human LH beta - 6%, Human CG alpha - 6%, Rat TSH - 4%. No reactivity was observed with human GH, prolactin, ACTH or mouse GH.
Follicle-stimulatine hormone (FSH) is a member of the glycoprotein hormone family. Members of this family consist of an identical alpha subunit and a hormone-specific beta subunit. Alternative splicing results in two transcript variants encoding the same protein. FSH is secreted by gonadotropic cells in the pituitary gland and in conjunction with luteinizing hormone, induces egg and sperm production.