FIGURE: 1 / 1
Essential cell division protein that catalyzes cross-linking of the peptidoglycan cell wall at the division septum (PubMed:1103132, PubMed:6450748, PubMed:9614966, PubMed:3531167, PubMed:7030331). Required for localization of FtsN (PubMed:9282742).
Protein Aliases: pbpB