Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
FSD1 encodes a centrosome associated protein that is characterized by an N-terminal coiled-coil region downstream of B-box (BBC) domain, a central fibronectin type III domain, and a C-terminal repeats in splA and RyR (SPRY) domain. The encoded protein associates with a subset of microtubules and may be involved in the stability and organization of microtubules during cytokinesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fibronectin type 3 and SPRY (spla, ryanodine) domain containing (with coiled-coil motif) 1; fibronectin type 3 and SPRY domain containing 1; fibronectin type 3 and SPRY domain-containing protein; Fibronectin type III and SPRY domain-containing protein 1; Microtubule-associated protein GLFND; MID1-related protein 1; midline 1-related protein 1
Gene Aliases: FSD1; GLFND; MIR1; VLP27
UniProt ID: (Human) Q9BTV5, (Mouse) Q7TPM6
Entrez Gene ID: (Human) 79187, (Mouse) 240121, (Rat) 301506
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support