Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGDERTAVSL VTALPGRPSS CVSVTDGPKF EVKLKKNANG LGFSFVQMEK ESCSHLKSDL VRIKRLFPGQ PAEENGAIAA GDIILAVNGR STEGLIFQEV LHLLRGAPQE VTLLLCRPPP GALPELEQEW QTPELSADKE FTRATCTDSC TSPILDQEDS WRDSASPDAG EGLGLRPESS QKAIREAQWG QNRERPWASS LTHSPESHPH LCKLHQERDE STLATSLEKD VRQNCYSVCD IMRLGRYSFS SPLTRLSTDI F
The FERM and PDZ domain containing (FRMPD) protein family consists of four proteins that contain a FERM (Four-point-one, erzin, radixin, moesin) domain and at least one PDZ (PSD-95/Discs large/Zonula-occuldens-1) domain. FRMPD2 also contains an N-terminal KIND domain and three PDZ domains and is structurally similar to the protein tyrosine phosphatase PTP-BL. FRMPD2 is localized in a polarized fashion in epithelial cells at the basolateral membrane and partially co-localizes with the tight-junction marker protein Zonula-occuldens-1. Suppression of FRMPD2 expression via RNAi in Caco-2 cells results in an impairment of tight junction formation, indicating that FRMPD2 plays a major role in tight junction formation. Other experiments indicate that FRMPD2 is a binding partner to several catenin family members and recruitment of FRMPD2 to cell-cell contacts is dependent on E-cadherin-mediated cell-cell adhesion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FERM and PDZ domain-containing protein 2; PDZ domain containing 5C; PDZ domain-containing protein 4; PDZ domain-containing protein 5C
Gene Aliases: FRMPD2; PDZD5C; PDZK4; PDZK5C
UniProt ID: (Human) Q68DX3
Entrez Gene ID: (Human) 143162
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support