Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAEYSYVKST KLVLKGTKTK SKKKKSKDKK RKREEDEETQ LDIVGIWWTV TNFGEISGTI AIEMDKGTYI HALDNGLFTL GAPHKEVDEG PSPPEQFTAV KLSDSRIALK SGYGKYLGIN SDGLVVGRSD AIGPREQWEP VFQNGKMALL ASNSCFIRCN EAGDIEAKSK TAGEEEMIKI RSCAERETKK KDDIPEEDKG NVKQCEINYV KKFQSFQDHK LKISKEDSKI LKKARKDGFL HETLLDRRAK LKADRYCK
This gene maps to a location 100 kb centromeric of the repeat units on chromosome 4q35 which are deleted in facioscapulohumeral muscular dystrophy . It is evolutionarily conserved and has related sequences on multiple human chromosomes but DNA sequence analysis did not reveal any homology to known genes. In vivo studies demonstrate the encoded protein is localized to the nucleolus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: facioscapulohumeral muscular dystrophy region gene-1; FSHD region gene 1 protein; Protein FRG1
Gene Aliases: FRG1; FRG1A; FSG1
UniProt ID: (Human) Q14331
Entrez Gene ID: (Human) 2483
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support