PA1-31654 detects FOXG1 from human samples. Expected to cross react with rat and mouse due to sequence homology.
PA1-31654 has been successfully used in Western blot applications. A suggested positive control for this product is human brain lysate.
The PA1-31654 immunogen is: Peptide with sequence HQNQGSSSNPLIH, from C Terminus of the protein sequence according to NP_004462 (FOXG1A); NP_005240 (FOXG1B)
FOXG1B belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of FOXG1B has not yet been determined; however, it may play a role in the development of the brain and telencephalonThis gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon.This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon.