Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • POMK Antibodies

          Abnova

          FLJ23356 Polyclonal Antibody, MaxPab™

          View all (25) POMK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite FLJ23356 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (2)
          FLJ23356 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FLJ23356 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FLJ23356 Antibody (H00084197-D01P) in WB

          Western Blot analysis of SGK196 expression in transfected 293T cell line (H00084197-T01) by SGK196 MaxPab polyclonal antibody.

          Lane 1: FLJ23356 transfected lysate(40.00 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FLJ23356 Antibody in Western Blot (WB)
          FLJ23356 Antibody in Western Blot (WB)

          Product Details

          H00084197-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          FLJ23356 (NP_115613.1, 1 a.a. approximately 350 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MEKQPQNSRR GLAPREVPPA VGLLLIMALM NTLLYLCLDH FFIAPRQSTV DPTHCPYGHF RIGQMKNCSP WLSCEELRTE VRQLKRVGEG AVKRVFLSEW KEHKVALSQL TSLEMKDDFL HGLQMLKSLQ GTHVVTLLGY CEDDNTMLTE YHPLGSLSNL EETLNLSKYQ NVNTWQHRLE LAMDYVSIIN YLHHSPVGTR VMCDSNDLPK TLSQYLLTSN FSILANDLDA LPLVNHSSGM LVKCGHRELH GDFVAPEQLW PYGEDVPFHD DLMPSYDEKI DIWKIPDISS FLLGHIEGSD MVRFHLFDIH KACKSQTPSE RPTAQDVLET YQKVLDTLRD AMMSQAREML

          Target Information

          This gene encodes a protein that may be involved in the presentation of the laminin-binding O-linked carbohydrate chain of alpha-dystroglycan (a-DG), which forms transmembrane linkages between the extracellular matrix and the exoskeleton. Some pathogens use this O-linked carbohydrate unit for host entry. Loss of function compound heterozygous mutations in this gene were found in a human patient affected by the Walker-Warburg syndrome (WWS) phenotype. Mice lacking this gene contain misplaced neurons (heterotopia) in some regions of the brain, possibly from defects in neuronal migration. Alternative splicing of this gene results in multiple transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: POMK; probable inactive protein kinase-like protein SgK196; Protein kinase-like protein SgK196; Protein O-mannose kinase; Sugen kinase 196

          View more View less

          Gene Aliases: 4930444A02Rik; MDDGA12; MDDGC12; POMK; SGK196

          View more View less

          UniProt ID: (Human) Q9H5K3, (Mouse) Q3TUA9

          View more View less

          Entrez Gene ID: (Human) 84197, (Mouse) 74653

          View more View less

          Function(s)
          protein kinase activity ATP binding phosphotransferase activity, alcohol group as acceptor carbohydrate kinase activity nucleotide binding phosphofructokinase activity phosphorylase kinase regulator activity cobinamide kinase activity phytol kinase activity kinase activity phosphatidylinositol phosphate kinase activity cyclin-dependent protein serine/threonine kinase regulator activity transferase activity transferase activity, transferring phosphorus-containing groups phenol kinase activity cyclin-dependent protein kinase activating kinase regulator activity inositol tetrakisphosphate 2-kinase activity heptose 7-phosphate kinase activity aminoglycoside phosphotransferase activity phosphatidylinositol 3-kinase activity eukaryotic elongation factor-2 kinase regulator activity eukaryotic elongation factor-2 kinase activator activity LPPG:FO 2-phospho-L-lactate transferase activity cytidine kinase activity glycerate 2-kinase activity (S)-lactate 2-kinase activity phosphoserine:homoserine phosphotransferase activity L-seryl-tRNA(Sec) kinase activity phosphocholine transferase activity polynucleotide 5'-hydroxyl-kinase activity ATP-dependent polynucleotide kinase activity GTP-dependent polynucleotide kinase activity farnesol kinase activity CTP:2-trans,-6-trans-farnesol kinase activity geraniol kinase activity geranylgeraniol kinase activity CTP:geranylgeraniol kinase activity prenol kinase activity 1-phosphatidylinositol-5-kinase activity 1-phosphatidylinositol-3-phosphate 4-kinase activity phosphatidylinositol-3,4-bisphosphate 5-kinase activity phosphatidylinositol bisphosphate kinase activity inositol-3,4,6-trisphosphate 1-kinase activity inositol 5-diphosphate pentakisphosphate 5-kinase activity inositol diphosphate tetrakisphosphate kinase activity
          Process(es)
          neuron migration protein phosphorylation protein O-linked glycosylation brain development learning or memory sensory perception of pain carbohydrate phosphorylation neuromuscular process phosphorylation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-588f58b695-w7k2p:80/100.66.134.75:80.
          git-commit: b8205085541c8e5054bc4cc0f27ee9f88e66ec64
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0