Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWTPELAILR GFPTEAERQQ WKQEGVVGSE SGSFLQLLLE GNYEAIFLNS MTQNIFNSTT TAEEKIDSYL EKQVVTFLDY STDLDTTERQ QLIFLLGVSS LQLFVQSNWT GPPVDLHPQD FLSSVLFQQF SEVKGLDAFV LSLLTLDGES IYSLTSKPIL LLLARIILVN VRHKLTAIQS LPWWTLRCVN IHQHLLEERS PLLFTLAENC IDQVMKLQNL FVDDSGRYLA IQFHLECAYV FLYYYEYRKA KDQLDIAKDI SQLQIDLTGA LGKRTRFQEN YVAQLILDVR REGDVLSNCE FTPAPTPQEH LTKNLELNDD TILNDIKLAD CEQFQMPDLC AEEIAIILGI CTNFQKNNPV HTLTEVELLA FTSCLLSQPK FWAIQTSALI LRTKLEKGST RRVERAMRQT QALADQFEDK TTSVLERLKI FYCCQVPPHW AIQRQLASLL FELGCTSSAL QIFEKLEMWE DVVICYERAG QHGKAEEILR QELEKKETPS LYCLLGDVLG DHSCYDKAWE LSRYRSARAQ RSKALLHLRN KEFQECVECF ERSVKINPMQ LGVWFSLGCA YLALEDYQGS AKAFQRCVTL EPDNAEAWNN LSTSYIRLKQ KVKAFRTLQE ALKCNYEHWQ IWENYILTST DVGEFSEAIK AYHRLLDLRD KYKDVQVLKI LVRAVIDGMT DRSGDVATGL KGKLQELFGR VTSRVTNDGE IWRLYAHVYG NGQSEKPDEN EKAFQCLSKA YKCDTQSNCW EKDITSFKEV VQRALGLAHV AIKCSKNKSS SQEAVQMLSS VRLNLRGLLS KAKQLFTDVA TGEMSRELAD DITAMDTLVT ELQDLSNQFR NQY
TTC27 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tetratricopeptide repeat protein 27; TPR repeat protein 27
Gene Aliases: TTC27
UniProt ID: (Human) Q6P3X3
Entrez Gene ID: (Human) 55622
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support