Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQQQPLPGPG APTTEPTKPP YSYIALIAMA IQSSPGQRAT LSGIYRYIMG RFAFYRHNRP GWQNSIRHNL SLNECFVKVP RDDRKPGKGS YWTLDPDCHD MFEHGSFLRR RRRFTRQTGA EGTRGPAKAR RGPLRATSQD PGVPNATTGR QCSFPPELPD PKGLSFGGLV GAMPASMCPA TTDGRPRPPM EPKEISTPKP ACPGELPVAT SSSSCPAFGF PAGFSEAESF NKAPTPVLSP ESGIGSSYQC RLQALNFCMG ADPGLEHLLA SAAPSPAPPT PPGSLRAPLP LPTDHKEPWV AGGFPVQGGS GYPLGLTPCL YRTPGMFFFE
The forkhead family of transcription factors belongs to the winged helix class of DNA-binding proteins. The protein encoded by this intronless gene contains a forkhead domain and is found predominantly in aorta and kidney. The function of the encoded protein is unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Forkhead box protein S1; Forkhead-like 18 protein; forkhead-related activator 10; Forkhead-related transcription factor 10; forkhead-related transcription factor FREAC-10; FREAC-10
Gene Aliases: FKHL18; FOXS1; FREAC10
UniProt ID: (Human) O43638
Entrez Gene ID: (Human) 2307
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support