Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MKGASEEKL ASVSNLVTVF ENSRTPEAAP RGHRLEDVHH RPECRPPESP GPREKTNVGE AVGSEPRTVS RRYLNSLKNK LSSEAWRKSC QPVTLSGSGT QEPEKKIVQE LLETEQAYVA RLHL (1-123 aa encoded by BC023645)
Activates CDC42, a member of the Ras-like family of Rho- and Rac proteins, by exchanging bound GDP for free GTP. Activates JNK1 via CDC42 but not RAC1. Binds to phosphatidylinositol 4,5- bisphosphate, phosphatidylinositol 3,4,5-trisphosphate, phosphatidylinositol 5-monophosphate, phosphatidylinositol 4- monophosphate and phosphatidylinositol 3-monophosphate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGD1 family, member 2; FLJ00276 protein; FYVE, RhoGEF and PH domain-containing protein 2; Zinc finger FYVE domain-containing protein 4
Gene Aliases: FGD2; ZFYVE4
UniProt ID: (Human) Q7Z6J4
Entrez Gene ID: (Human) 221472
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support