Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MELSGATMAR GLAVLLVLFL HIKNLPAQAA DTCPEVKVVG LEGSDKLTIL RGCPGLPGAP GPKGEAGVIG ERGERGLPGA PGKAGPVGPK GDRGEKGMRG EKGDAGQSQS CATGPRNCKD LLDRGYFLSG WHTIYLPDCR PLTVLCDMDT DGGGWTVFQR RMDGSVDFYR DWAAYKQGFG SQLGEFWLGN DNIHALTAQG SSELRVDLVD FEGNHQFAKY KSFKVADEAE KYKLVLGAFV GGSAGNSLTG HNNNFFSTKD QDNDVSSSNC AEKFQGAWWY ADCHASNLNG LYLMGPHESY ANGINWSAAK GYKYSYKVSE MKVRPA
M-Ficolin (Ficolin-1) is a non-serum protein that is expressed in leukocytes and lung. M-Ficolin precipitates with mannose-binding lectin (MBL)-associated serine proteases (MASP)-1 and MASP-2, indicating that M-Ficolin forms complexes with MASP-1 and MASP-2, recognizes certain types of bacteria and subsequently activates the lectin pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collagen/fibrinogen domain-containing protein 1; Ficolin-1; Ficolin-A; Ficolin-alpha; M-ficolin
Gene Aliases: FCN1; FCNM
UniProt ID: (Human) O00602
Entrez Gene ID: (Human) 2219
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support