Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHLKPYWKLQ KKEHPPEVSR ETQRTPMNHQ KAVNDETCKA SHITSSVFPS ASLGKASSRK PFGILSPNVL CSMSGKSPVE SSLNVKTKKN APSATIHQGE EEGPLDIWAV VKPGNTKEKI AFFASHQCSN RIGSMKIKSS WDIDGRATKR RKKSGDLKKA KVQVERMREV NSRCYQPEPF ACGIEHCSVH YVSDSGDGVY AGRPLSVIQM VAFLEQRASA LLASCSKNCT NSPAIVRFSG QSRGVPAVSE SYSAPGACEE PTERGNLEVG EPQSEPVRVL DMVAKLESEC LKRQGQREPG SLSRNNSFRR NVGRVLLANS TQADEGKTKK GVLEAPDTQV NPVGSVSVDC GPSRADRCSP KEDQAWDGAS QDCPPLPAGV SFHIDSAELE PGSQTAVKNS NRYDVEMTDE LVGLPFSSHT YSQASELPTD AVDCMSRELV SLTSRNPDQR KESLCISITV SKVDKDQPSI LNSCEDPVPG MLFFLPPGQH LSDYSQLNES TTKESSEASQ LEDAAGGDSA SEEKSGSAEP FVLPASSVES TLPVLEASSW KKQVSHDFLE TRFKIQQLLE PQQYMAFLPH HIMVKIFRLL PTKSLVALKC TCCYFKFIIE YYNIRPADSR WVRDPRYRED PCKQCKKKYV KGDVSLCRWH PKPYCQALPY GPGYWMCCHR SQKGFPGCKL GLHDNHWVPA CHSFNRAIHK KAKGTEAEEE Y
Members of the F-box protein family, such as FBXO34, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F-box only protein 34; protein CGI-301
Gene Aliases: CGI-301; FBX34; FBXO34
UniProt ID: (Human) Q9NWN3
Entrez Gene ID: (Human) 55030
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support