Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAAAEERMA EEGGGGQGDG GSSLASGSTQ RQPPPPAPQH PQPGSQALPA PALAPDQLPQ NNTLVALPIV AIENILSFMS YDEISQLRLV CKRMDLVCQR MLNQGFLKVE RYHNLCQKQV KAQLPRRESE RRNHSLARHA DILAAVETRL SLLNMTFMKY VDSNLCCFIP GKVIDEIYRV LRYVNSTRAP QRAHEVLQEL RDISSMAMEY FDEKIVPILK RKLPGSDVSG RLMGSPPVPG PSAALTTMQL FSKQNPSRQE VTKLQQQVKT NGAGVTVLRR EISELRTKVQ EQQKQLQDQD QKLLEQTQII GEQNARLAEL ERKLREVMES AVGNSSGSGQ NEESPRKRKK ATEAIDSLRK SKRLRNRK
Members of the F-box protein family, such as FBXO28, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centromere protein 30; F-box only protein 28
Gene Aliases: CENP-30; Fbx28; FBXO28; KIAA0483
UniProt ID: (Human) Q9NVF7
Entrez Gene ID: (Human) 23219
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support