Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is expected detect both isoforms.
Immunogen sequence: MPSEILLKI FSYLDAVSLL CTGCVSRRFY HLANDNFIWI GIYSTAFSPA RSNWKFNSVE KIAMSMSFLS VQDKEAGYWK KEYITKQIAS VKAALADILK PVNPYTGLPV KTKEALRIFG LGWAIILKEK GGKEYIMEHV DLSINDTSVT VIWYGKKWPC LASLSTLDLC GMTPVFTDWY KTPTKHRLRW HSLIAKYNLS HLTISTMIGC DRLIRIFCLH PGLLVGVWKK EEELAFVMAN LHFHHLVERS TLGSATIPYE LPPHSPFLDD SPEYGLHGYQ LHVDLHSGGV FYLCGTFRNL FT (1-301 aa encoded by BC029579)
Members of the F-box protein family, such as FBXO15, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F-box only protein 15; MGC39671
Gene Aliases: AU019763; ecat3; FBX15; FBXO15
UniProt ID: (Human) Q8NCQ5
Entrez Gene ID: (Human) 201456, (Mouse) 50764
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support