PA1-31771 detects EZH1 from human samples, but is expected to cross-react with other species due to sequence homology.
PA1-31771 has been successfully used in Western blot applications.
The PA1-31771 immunogen is: Synthetic peptide: EIPNPPTSKCITY, corresponding to amino acids 2-14 of Human EZH 1.
EZH1 is a component of a noncanonical Polycomb repressive complex-2 (PRC2) that mediates methylation of histone H3 lys27 (H3K27) and functions in the maintenance of embryonic stem cell pluripotency and plasticity (Shen et al., 2008 ).