Predicted reactivity: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EVX1 is a member of the vertebrate eve-related homeo box family. This protein acts as a transcriptional repressor. A similar protein in mice is critical for early embryogenesis.