Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: GPPSSRPSSR VVNGEDAVPY SWPWQVSLQY EKSGSFYHTC GGSLIAPDWV VTAGHCISSS WTYQVVLGEY DRAVKEGPEQ VIPINSGDLF VHPLWNRSCV ACGNDIALIK LSRSAQLGDA VQLASLPPAG DILPNETPCY ITGWGRLYTN GPLPDKLQEA LLPVVDYEHC SRWNWWGSSV KKTMVCAGGD IRSGCNGDSG GPLNCPTEDG GWQVHGVTSF VSAFGCNTRR KPTVFTRVSA FIDWIEETIA SH
Efficient protease with alanine specificity but only little elastolytic activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cholesterol-binding pancreatic protease; Chymotrypsin-like elastase family member 3B; elastase 3B, pancreatic; Elastase IIIB; Elastase-3B; fecal elastase 1; pancreatic elastase 1; pancreatic endopeptidase E; Protease E; proteinase E
Gene Aliases: CBPP; CELA3B; ELA3B
UniProt ID: (Human) P08861
Entrez Gene ID: (Human) 23436
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support