Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • NFIL3 (E4BP4) Antibodies

          Invitrogen

          E4BP4 (NFIL3) Monoclonal Antibody (MABA223), PE, eBioscience™

          2 Published Figures
          2 References
          View all (15) NFIL3 (E4BP4) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite E4BP4 (NFIL3) Monoclonal Antibody (MABA223), PE, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (2)
          E4BP4 (NFIL3) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          E4BP4 (NFIL3) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          E4BP4 (NFIL3) Antibody (12-9812-42) in Flow

          Normal human peripheral blood cells were unstimulated (left) or stimulated with Anti-Human CD3 Functional Grade Purified (Product # 16-0037-81) and Anti-Human CD28 Functional Grade Purified (Product # 16-0289-81) overnight (right) and then fixed and permeabilized with the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00). Cells were intracellularly stained with Anti-Human CD20 FITC (Product # 11-0209-42) and Anti-Human E4BP4 (NFIL3) PE. Viable cells, as determined by staining with Fixable Viability Dye eFluor® 780 (Prod... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          E4BP4 (NFIL3) Antibody in Flow Cytometry (Flow)
          E4BP4 (NFIL3) Antibody in Flow Cytometry (Flow)
          E4BP4 (NFIL3) Antibody in Flow Cytometry (Flow)

          Product Details

          12-9812-42

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          5 µL (0.125 µg)/test
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Recommended Isotype Control

          Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), PE, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          MABA223

          Immunogen

          21H2-hNFIL3 (aa 163-462)
          View immunogen

          Conjugate

          PE PE PE
          • Alexa Fluor 647 
          • Request custom conjugation

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Liquid

          Concentration

          5 µL/Test

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 0.2% BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11042477

          Product Specific Information

          Description: This MABA223 monoclonal antibody recognizes human E4BP4 (NFIL3), a basic leucine zipper transcription factor first identified for its binding to the promoters of the adenovirus E4 protein and human IL-3. E4BP4 is involved in the transcriptional control of the mammalian circadian clock, motor neuron growth and survival, regulation of von Willbrand factor expression, and osteoblast function. In addition, it is required for the development of NK and NKT cells as well as CD8a+ dendritic cells. Although relatively high levels of mRNA are reported in resting NK and NKT cells, studies at eBioscience have observed expression of E4BP4 protein in lymphocytes only after activation.

          Applications Reported: This MABA223 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

          Applications Tested: This MABA223 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00). This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

          Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Expression of interleukin-3 (IL3; MIM 147740) is restricted to activated T cells, natural killer (NK) cells, and mast cell lines. Transcription initiation depends on the activating capacity of specific protein factors, such as NFIL3, that bind to regulatory regions of the gene, usually upstream of the transcription start site (Zhang et al., 1995 [PubMed 7565758]).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: E4 promoter-binding protein 4; Interleukin-3 promoter transcriptional activator; Interleukin-3-binding protein 1; NF IL3A; nuclear factor interleukin 3 regulated protein; Nuclear factor interleukin-3-regulated protein; OTTHUMP00000021633; Transcriptional activator NF-IL3A

          View more View less

          Gene Aliases: E4BP4; IL3BP1; NF-IL3A; NFIL3; NFIL3A

          View more View less

          UniProt ID: (Human) Q16649

          View more View less

          Entrez Gene ID: (Human) 4783

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding transcription corepressor activity protein binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter immune response circadian rhythm positive regulation of gene expression cellular response to interleukin-4
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7dfb8678b5-vqhv7:80/100.66.129.46:80.
          git-commit: 632cddff25488e9c1007a93c87c1899f725eef0d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0