Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Desmoplakin Antibodies

          Progen

          Desmoplakin 1/2 Antibody Cocktail

          View all (24) Desmoplakin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Desmoplakin 1/2 Antibody Cocktail

          Product Details

          651109

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          Ready-to-use
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Ready-to-use
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse

          Host/Isotype

          Mouse / IgG2b

          Class

          Cocktail

          Type

          Antibody

          Clone

          DP I/II 236.23.1

          Immunogen

          C-Terminal Polypeptide Of Recombinant Human Desmoplakin (Starting At Amino Acid Position 1980)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          651109 is a mixture of monoclonal antibodies that target Desmoplakin 1/2.

          Specificity: distinct punctate membrane staining of different epithelia. Desmosomes of the myocardium are also positive. Tumors specifically detected: Primary and metastatic carcinoma and meningioma. Polypeptides reacting: Desmoplakin 1 and 2 (MW 250 000 and 215 000)Reactivity on cultured cell lines: Several human carcinoma cell lines: MCF-7,A-431, HaCaT, CaCo2; bovine cells: MDBK, BMGE.

          Target Information

          Desmoplakin (DSP) is a major high molecular weight protein of the desmosomes. It is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes. Desmoplakin mutations are associated with Keratoderma and cardiomyopathy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 250/210 kDa paraneoplastic pemphigus antigen; Desmoplakin; desmoplakin I/II; Desmoplakin II; Desmoplakin-I; desmosomal cytoskeletal connector molecule; DP

          View more View less

          Gene Aliases: 2300002E22Rik; 5730453H04Rik; AA407887; AA407888; AW109828; BOS_22012; DCWHKTA; DP; DSP; rul

          View more View less

          UniProt ID: (Human) P15924, (Mouse) E9Q557

          View more View less

          Entrez Gene ID: (Human) 1832, (Bovine) 514360, (Mouse) 109620

          View more View less

          Function(s)
          protein kinase C binding structural molecule activity structural constituent of cytoskeleton protein binding protein binding, bridging poly(A) RNA binding cell adhesion molecule binding cell adhesive protein binding involved in bundle of His cell-Purkinje myocyte communication scaffold protein binding
          Process(es)
          desmosome organization ventricular compact myocardium morphogenesis epidermis development peptide cross-linking keratinocyte differentiation adherens junction organization wound healing intermediate filament organization protein localization to adherens junction bundle of His cell-Purkinje myocyte adhesion involved in cell communication regulation of heart rate by cardiac conduction regulation of ventricular cardiac muscle cell action potential single organismal cell-cell adhesion skin development intermediate filament cytoskeleton organization
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5dd894dbdc-64gxr:80/100.66.134.75:80.
          git-commit: 83227db6267f00a4ced007f33c273793adf466ab
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0