Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Gene ontology (GO) annotation includes protein C-linked glycosylation via 2'-alpha-mannosyl-L-tryptophan; spermatid development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dpy-19-like 2; dpy-19-like 2 (C. elegans); Dpy-19-like protein 2; Probable C-mannosyltransferase DPY19L2; Protein dpy-19 homolog 2; spermatogenesis associated 34
Gene Aliases: 4932443J21Rik; DPY19L2; Dyp19l2; Gm18376; RGD1564311; SPATA34; SPGF9; UNQ3127/PRO10284
UniProt ID: (Human) A4FVC1, (Mouse) P0CW70
Entrez Gene ID: (Human) 283417, (Rat) 300461, (Mouse) 320752
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support