Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: FDRDLTLAG RVIVKCPTSG RLEQQLVHVE RPILRIPHLA IHLQRNINEN FGPNTEMHLV PILATAIQEE LEKGTPEPGP LNAVDERHHS VLMSLLCAHL GLSPKDIVEM ELCLADTQPA VLGGAYDEFI FAPRLDNLHS CFCALQALID SCAGPGSLAT EPHVRMVTLY DNEEVGSESA QGAQSLLTEL VLRRISASCQ HPTAFEEAIP KSFMISADMA HAVHPNYLDK HEENHRPLFH KGPVIKVNSK QRYASNAVSE ALIREVANKV KVPLQDLMVR NDTPCGTTIG PILASRLGLR VLDLGSPQLA MHSIREMACT TGVLQTLTLF KGFFELFPSL SHNLLVD (130-475 aa encoded by BC000653 )
Aminopeptidase with specificity towards an acidic amino acid at the N-terminus. Likely to play an important role in intracellular protein and peptide metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aspartyl aminopeptidase
Gene Aliases: ASPEP; DAP; DNPEP
UniProt ID: (Human) Q9ULA0
Entrez Gene ID: (Human) 23549
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support