FIGURE: 1 / 1
This antibody has been tested in direct-ELISA
Protects DNA from oxidative damage by sequestering intracellular Fe(2+) ion and storing it in the form of Fe(3+) oxyhydroxide mineral. One hydrogen peroxide oxidizes two Fe(2+) ions, which prevents hydroxyl radical production by the Fenton reaction. It protects DNA from hydroxyl radical-mediated cleavage. Binds DNA with no apparent sequence specificity without self-aggregation nor promotion of DNA condensation. Is unable to protect DNA from DNase-mediated cleavage.