Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: NKANPRCPES LTRDTATEIA INVKALYNET ESLLVGRVPL QLESPHEERV SNALHSVEVE LQKLTEIPWL YYILHPNEDE EPPMDCTKRN NRS
Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
This gene encodes a member of the diacylglycerol kinase (DGK) enzyme family. Members of this family are involved in regulating intracellular concentrations of diacylglycerol and phosphatidic acid. Variation in this gene has been associated with bipolar disorder. Alternatively spliced transcript variants have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAG kinase eta; DGK-eta; Diacylglycerol kinase eta; Diglyceride kinase eta; DKFZp761I1510; EC 2.7.1.107
Gene Aliases: DGKeta; DGKH
UniProt ID: (Human) Q86XP1
Entrez Gene ID: (Human) 160851
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support