Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Sequence of this protein is as follows: MALRKELLKS IWYAFTALDV EKSGKVSKSQ LKVLSHNLYT VLHIPHDPVA LEEHFRDDDD GPVSSQGYMP YLNKYILDKV EEGAFVKEHF DELCWTLTAK KNYRADSNGN SMLSNQDAFR LWCLFNFLSE DKYPLIMVPD EVEYLLKKVL SSMSLEVSLG ELEELLAQEA QVAQTTGGLS VWQFLELFNS GRCLRGVGRD TLSMAIHEVY QELIQDVLKQ GYLWKRGHLR RNWAERWFQL QPSCLCYFGS EECKEKRGII PLDAHCCVEV LPDRDGKRCM FCVKTATRTY EMSASDTRQR QEWTAAIQMA IRLQAEGKTS LHKDLKQKRR EQREQRERRR AAKEEELLRL QQLQEEKERK LQELELLQEA QRQAERLLQE EEERRRSQHR ELQQALEGQL REAEQARASM QAEMELKEEE AARQRQRIKE LEEMQQRLQE ALQLEVKARR DEESVRIAQT RLLEEEEEKL KQLMQLKEEQ ERYIERAQQE KEELQQEMAQ QSRSLQQAQQ QLEEVRQNRQ RADEDVEAAQ RKLRQASTNV KHWNVQMNRL MHPIEPGDKR PVTSSSFSGF QPPLLAHRDS SLKRLTRWGS QGNRTPSPNS NEQQKSLNGG DEAPAPASTP QEDKLDPAPE N
DEF6, or IBP, is a guanine nucleotide exchange factor for RAC and CDC42 that is highly expressed in B and T cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEF-6; DEF6 guanine nucleotide exchange factor; Differentially expressed in FDCP 6 homolog; IRF4-binding protein; OTTHUMP00000016251; RP3-329A5.5; SWAP-70-like adaptor protein of T cells
Gene Aliases: DEF6; IBP; SLAT; SWAP70L
UniProt ID: (Human) Q9H4E7
Entrez Gene ID: (Human) 50619
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support